missing translation for 'onlineSavingsMsg'
Learn More

Pipamazine, TRC

Product Code. 17962885
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962885 1 g 1g
17913185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17962885 Supplier Toronto Research Chemicals Supplier No. TRCP4751901G
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-04-8
Molecular Formula C21 H24 Cl N3 O S
Molecular Weight (g/mol) 401.95
Synonym 4-Piperidinecarboxamide, 1-[3-(2-chloro-10H-phenothiazin-10-yl)propyl]-Isonipecotamide, 1-[3-(2-chlorophenothiazin-10-yl)propyl]- (6CI,7CI,8CI), 1-[3-(2-Chlorophenothiazin-10-yl)propyl]isonipecotamide, 10-[3-(4-Carbamoylpiperidino)propyl]-2-chlorophenothiazine, Mornidine, Nausidol, Nometine, Pipamazine, RP 9153, SC 9387
IUPAC Name 1-[3-(2-chlorophenothiazin-10-yl)propyl]piperidine-4-carboxamide
SMILES NC(=O)C1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H24ClN3OS/c22-16-6-7-20-18(14-16)25(17-4-1-2-5-19(17)27-20)11-3-10-24-12-8-15(9-13-24)21(23)26/h1-2,4-7,14-15H,3,8-13H2,(H2,23,26)
Formula Weight 401.1329
Chemical Name or Material Pipamazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.