missing translation for 'onlineSavingsMsg'
Learn More

Pipamazine, LoGiCal

Product Code. 30459267
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459267 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459267 Supplier LoGiCal Supplier No. LGCFOR1603.00
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pipamazine
CAS 84-04-8
Analyte or Component Names Pipamazine
Shipping Condition Room Temperature
Molecular Formula C21 H24 Cl N3 O S
Certifications/Compliance ISO 9001
Synonym 4-Piperidinecarboxamide, 1-[3-(2-chloro-10H-phenothiazin-10-yl)propyl]-Isonipecotamide, 1-[3-(2-chlorophenothiazin-10-yl)propyl]- (6CI,7CI,8CI), 1-[3-(2-Chlorophenothiazin-10-yl)propyl]isonipecotamide, 10-[3-(4-Carbamoylpiperidino)propyl]-2-chlorophenothiazine, Mornidine, Nausidol, Nometine, Pipamazine, RP 9153, SC 9387
InChI Formula InChI=1S/C21H24ClN3OS/c22-16-6-7-20-18(14-16)25(17-4-1-2-5-19(17)27-20)11-3-10-24-12-8-15(9-13-24)21(23)26/h1-2,4-7,14-15H,3,8-13H2,(H2,23,26)
Solution Type Neat
SMILES NC(=O)C1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1
IUPAC Name 1-[3-(2-chlorophenothiazin-10-yl)propyl]piperidine-4-carboxamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 401.95
Quantity 5 mg
Formula Weight 401.1329 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.