missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP5K2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19837
This item is not returnable.
View return policy
Description
PIP5K2B Polyclonal specifically detects PIP5K2B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PIP5K2B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, EC 2.7.1, EC 2.7.1.149, phosphatidylinositol-4-phosphate 5-kinase, type II, beta, Phosphatidylinositol-5-phosphate 4-kinase type II beta, phosphatidylinositol-5-phosphate 4-kinase type-2 beta, phosphatidylinositol-5-phosphate 4-kinase, type II, beta, PI(5)P 4-kinase type II beta, PI5P4KB, PIP4KII-beta, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PTDINS(4)P-5-kinase, PtdIns(5)P-4-kinase isoform 2-beta | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Cancer | |
| 8396 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.83mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P78356 | |
| PIP4K2B | |
| Recombinant protein encompassing a sequence within the center region of human PIP4K2B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (100%), Rat (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction