missing translation for 'onlineSavingsMsg'
Learn More

PIP5K2 alpha Antibody (OTI3D3), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18426308 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426308 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426308 Supplier Novus Biologicals Supplier No. NBP273417MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIP5K2 alpha Monoclonal antibody specifically detects PIP5K2 alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K2 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3D3
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-4-phosphate-5-kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha, Diphosphoinositide kinase 2-alpha, EC 2.7.1, EC 2.7.1.149, FLJ13267, phosphatidylinositol-4-phosphate 5-kinase, type II, alpha, Phosphatidylinositol-5-phosphate 4-kinase type II alpha, phosphatidylinositol-5-phosphate 4-kinase type-2 alpha, phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, PI(5)P 4-kinase type II alpha, PIP4KII-alpha, PIP5K2, PIP5K2API5P4KA, PIP5KIIA, PIP5KIIalpha, PIP5KII-alpha, PIP5KIII, PIPK, PtdIns(4)P-5-kinase B isoform, PtdIns(4)P-5-kinase C isoform, PtdIns(5)P-4-kinase isoform 2-alpha, type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Host Species Mouse
Immunogen Full length human recombinant protein of human PIP4K2A (NP_005019) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5305
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.