missing translation for 'onlineSavingsMsg'
Learn More

PIP5K2 alpha Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30763099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763099 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763099 Supplier Novus Biologicals Supplier No. NBP347141MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIP5K2 alpha Polyclonal antibody specifically detects PIP5K2 alpha in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K2 alpha
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-4-phosphate-5-kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha, Diphosphoinositide kinase 2-alpha, EC 2.7.1, EC 2.7.1.149, FLJ13267, phosphatidylinositol-4-phosphate 5-kinase, type II, alpha, Phosphatidylinositol-5-phosphate 4-kinase type II alpha, phosphatidylinositol-5-phosphate 4-kinase type-2 alpha, phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, PI(5)P 4-kinase type II alpha, PIP4KII-alpha, PIP5K2, PIP5K2API5P4KA, PIP5KIIA, PIP5KIIalpha, PIP5KII-alpha, PIP5KIII, PIPK, PtdIns(4)P-5-kinase B isoform, PtdIns(4)P-5-kinase C isoform, PtdIns(5)P-4-kinase isoform 2-alpha, type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Host Species Rabbit
Immunogen phosphatidylinositol-5-phosphate 4-kinase, type II, alpha. (Uniprot# P48426)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5305
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.