missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIP5K1C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575549
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PIP5K1C lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 gamma phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1C is found at synapses and has been found to play roles in endocytosis and cell migration. PIP5K1C directly associates with -beta-catenin and increases -beta-catenin activity downstream of growth factor stimulation. PIP5K1C expression and kinase activity enhance -beta-catenin phosphorylation on residues that promote nuclear importation and transcriptional activity. PIP5K1C deficiency impairs neutrophil recruitment as a result of defect in adhesion. PIP5K1C regulated the adhesion through facilitation of RhoA GTPase and integrin activation by chemo attractants.
Specifications
| PIP5K1C | |
| Polyclonal | |
| Unconjugated | |
| PIP5K1C | |
| AI115456; AI835305; diphosphoinositide kinase; Kiaa0589; LCCS3; phosphatidylinositol 4-phosphate 5-kinase type I gamma; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type 1 gamma; phosphatidylinositol-4-phosphate 5-kinase type I gamma; phosphatidylinositol-4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; Pip5k1c; PIP5K1-gamma; PIP5Kgamma; PIP5K-GAMMA; PIP5KI gamma; Pip5kIgamma; PIP5KI-gamma; PIPKIg_v4; ptdIns(4)P-5-kinase 1 gamma; ptdIns(4)P-5-kinase gamma; ptdInsPKIgamma; type I PIP kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18717, 23396, 314641 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60331, O70161, Q5I6B8 | |
| PIP5K1C | |
| Synthetic peptide corresponding to amino acids 346-391 of Human PIP5K1C. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction