missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIP5K1 alpha Polyclonal specifically detects PIP5K1 alpha in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
Specifications
| Antigen | PIP5K1 alpha |
| Applications | Western Blot, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha, EC 2.7.1, EC 2.7.1.68, Phosphatidylinositol-4-phosphate 5-kinase type I alpha, phosphatidylinositol-4-phosphate 5-kinase type-1 alpha, phosphatidylinositol-4-phosphate 5-kinase, type I, alpha, PIP5K1-alpha, PIP5KIalpha, PtdIns(4)P-5-kinase 1 alpha |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 231-500 of human PIP5K1 alpha (NP_001129109.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?