missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIP4K2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566471
This item is not returnable.
View return policy
Description
Immunogen sequence: PDSPGNLLSFP RFFGPGEFDP SVDVYAMKSH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
Specifications
| PIP4K2B | |
| Polyclonal | |
| Unconjugated | |
| PIP4K2B | |
| 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; AI848124; c11; diphosphoinositide kinase 2-beta; LOW QUALITY PROTEIN: phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type 2 beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP4KII-beta; Pip5k2b; PIP5KIIB; PIP5KIIbeta; PIP5P4KB; PIPKIIgamma; PTDINS(4)P-5-kinase; ptdIns(5)P-4-kinase isoform 2-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8396 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78356 | |
| PIP4K2B | |
| Recombinant Human PIP4K2B. Recombinant protein control fragment (Product #RP-105227). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction