missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PIP2:PS Lipid Kinase Substrate

Product Code. 10627805
Change view
Click to view available options
Quantity:
400 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10627805 400 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10627805 Supplier Thermo Scientific™ Supplier No. PV5100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical ADP accumulation
Chromagen Quantity 400nmol
Content And Storage PIP2:PS Lipid Kinase Substrate in 50 mM HEPES pH 7.5, 3 mM MgCl2, and 1 mM EGTA.

Store at -20°C.
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation, Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), Adapta™ Universal Kinase Assay (PV5099), drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity
Quantity 400 nmol
Substrate Phosphoinositol-4,5-bisphosphate (PIP2) lipid vesicles with phosphotidylserine (PS)
Substrate Properties Lipid-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Lipid Kinases
Gene ID (Entrez) 5290, 5294
Item Description PIP2:PS Lipid Kinase Substrate, 400nmol, consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphatidylserine (PS)
Product Type PIP2:PS Lipid Kinase Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry PIK3CG, PIK3CD/PIK3R1, PIK3CA/PIK3R1
Validated Kinases PIK3CA, PIK3CG, PIK3CB, PIK3CD
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.