missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Mouse anti-Human, Clone-PIP/1571, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54451-100UG
This item is not returnable.
View return policy
Description
PIP Monoclonal specifically detects PIP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.
Specifications
| PIP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| Mouse | |
| 15 kDa | |
| 100 μg | |
| Cancer, Cellular Markers, Signal Transduction | |
| 5304 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| PIP/1571 | |
| Unconjugated | |
| PBS with No Preservative | |
| PIP | |
| Recombinant human GCDFP-15 protein fragment (aa 41-146) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction