missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£289.00 - £428.00
Specifications
| Antigen | PIP |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Proximity Ligation Assay |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18410341
|
Novus Biologicals
NBP1-83386-25ul |
25 μL |
£289.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18089913
|
Novus Biologicals
NBP1-83386 |
0.1 mL |
£428.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIP Polyclonal specifically detects PIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Proximity Ligation Assay.Specifications
| PIP | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay | |
| Unconjugated | |
| RUO | |
| Human | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| PIP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Proximity Ligation Assay | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5304 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FDIPKSVRPNDEVTAVLAVQTELKECMVVKTYLISSIPLQGAFNYKYTACLCDDNPKTFYWDFYTNRTVQIAAVVDVIRELGICPDDAAVIPIKNNRFYTIEILKVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title