missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone, MedChemExpress

Product Code. p-200101025
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16405938

Brand: Medchem Express HY13956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pioglitazone (U 72107) is an orally active and selective PPARγ (peroxisome proliferator-activated receptor) agonist with high affinity binding to the PPARγ ligand-binding domain with EC50 of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. Pioglitazone can be used in diabetes research.

Pioglitazone (U 72107) is an orally active and selective PPARγ (peroxisome proliferator-activated receptor) agonist with high affinity binding to the PPARγ ligand-binding domain with EC50 of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. Pioglitazone can be used in diabetes research.

Specifications

Chemical Name or Material Pioglitazone
CAS 111025-46-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20N2O3S
Quantity 10 mg
Synonym U 72107
Solubility Information DMSO : 40 mg/mL (112.22 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N1)SC(CC2=CC=C(OCCC3=NC=C(CC)C=C3)C=C2)C1=O
Molecular Weight (g/mol) 356.44
Formula Weight 356.44
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.