missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pioglitazone hydrochloride

Product Code. 18055732
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055732 50 mg 50mg
18054252 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055732 Supplier Tocris Bioscience™ Supplier No. 4124/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective PPARγ agonist; antidiabetic agent

Pioglitazone hydrochloride is a selective PPARγ agonist (EC50 = 0.69 μM). Thiazolidinedione (TZD) derivative and antidiabetic agent; improves insulin sensitivity. Also enhances proliferation and survival of mESCs and acts synergistically with Y-27632 (Cat. No. 1254) to improve survival of dissociated single hPSCs and increase colony formation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112529-15-4
Molecular Formula C19H21ClN2O3S

Specifications

Quantity 50 mg
Formula Weight 392.9
Purity >99%
Molecular Weight (g/mol) 392.898
Chemical Name or Material Pioglitazone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.