missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pioglitazone hydrochloride
Description
Pioglitazone hydrochloride is a selective PPARγ agonist (EC50 = 0.69 μM). Thiazolidinedione (TZD) derivative and antidiabetic agent; improves insulin sensitivity. Also enhances proliferation and survival of mESCs and acts synergistically with Y-27632 (Cat. No. 1254) to improve survival of dissociated single hPSCs and increase colony formation.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 392.9 |
| Purity | >99% |
| Molecular Weight (g/mol) | 392.898 |
| Chemical Name or Material | Pioglitazone hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?