missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone hydrochloride, MedChemExpress

Product Code. p-200099415
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16491736

Brand: Medchem Express HY14601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pioglitazone hydrochloride is a potent and selective PPARγ agonist with EC50s of 0.93 and 0.99 μM for human and mouse PPARγ, respectively.

Pioglitazone hydrochloride is a potent and selective PPARγ agonist with EC50s of 0.93 and 0.99 μM for human and mouse PPARγ, respectively.

Specifications

Chemical Name or Material Pioglitazone hydrochloride
CAS 112529-15-4
Color White
Purity Grade Notes Research
Molecular Formula C19H21ClN2O3S
Quantity 500 mg
Synonym U 72107A AD 4833
Solubility Information DMSO : 100 mg/mL (254.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CCC1=CN=C(CCOC2=CC=C(CC3C(NC(S3)=O)=O)C=C2)C=C1.Cl
Molecular Weight (g/mol) 392.9
Formula Weight 392.9
Percent Purity 99.75%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.