missing translation for 'onlineSavingsMsg'
Learn More

Pinoresinol Diglucoside, MedChemExpress

Product Code. 16481468
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481468 5 mg 5mg
16444867 50 mg 50mg
16429058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481468 Supplier Medchem Express Supplier No. HYN0657
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.

Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.

Specifications

Chemical Name or Material Pinoresinol Diglucoside
CAS 63902-38-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H42O16
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (146.48 mM; Need ultrasonic)
SMILES COC(C=C1[C@@H]2[C@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C=C4)([H])[C@]3([H])CO2)=C(C=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO
Molecular Weight (g/mol) 682.67
Formula Weight 682.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.