missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£181.00 - £410.00
Specifications
| Antigen | PINK1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006355
|
Novus Biologicals
NB100-493SS |
0.025 mL |
£181.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18716263
|
Novus Biologicals
NB100-493 |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PINK1 Polyclonal specifically detects PINK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| PINK1 | |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial | |
| PINK1 | |
| IgG | |
| Affinity Purified | |
| PINK1 Antibody is expected recognize isoform 1 and basted on a homology blast, will not recognize isoform 2. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q9BXM7 | |
| 65018 | |
| PINK1 antibody was developed using an N-terminal region synthetic peptide made to the human PINK1 protein sequence (between residues 1-50). [UniProt# Q9BXM7] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 62.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title