missing translation for 'onlineSavingsMsg'
Learn More

PINK1 Antibody (420920) [DyLight 405], Novus Biologicals™

Product Code. 30735840 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735840 Supplier Novus Biologicals Supplier No. FAB4357E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

PINK1 Monoclonal antibody specifically detects PINK1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PINK1
Applications Immunohistochemistry
Classification Monoclonal
Clone 420920
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial
Host Species Rat
Immunogen E. coli-derived recombinant human PINK1, Met1-Leu581, Accession # ABM87645
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Autophagy, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 65018
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.