missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18266-1-AP-150UL
This item is not returnable.
View return policy
Description
TSC22D4 is a leucine zipper-containing protein that is highly conserved during evolution. It is transcriptionally up-regulated by many different stimuli, including anti-cancer drugs and growth inhibitors. TSC22D4 may play a suppressive role in tumorigenesis.Specifications
| Pinin | |
| Polyclonal | |
| Unconjugated | |
| PNN | |
| Desmosome associated protein, Domain rich serine protein, DRS, DRS protein, DRSP, memA, Nuclear protein SDK3, Pinin, PNN, SDK3, SR like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5411 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H307 | |
| PNN | |
| Pinin Fusion Protein Ag6643 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction