missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-55315
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Pinin Polyclonal specifically detects Pinin in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones
| Pinin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| 140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5411 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q9H307 | |
| PNN | |
| The immunogen recognized by this antibody maps to a region between residue 667 and the C-terminus (residue 717) of human Pinin, desmosome associated protein using the numbering given in entry NP_002678.2 (GeneID 5411). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido