missing translation for 'onlineSavingsMsg'
Learn More

Pinin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35710-20ul

 View more versions of this product

Product Code. 30230860

  • £145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pinin Polyclonal antibody specifically detects Pinin in Human,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pinin
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Pinin (NP_002678.2).,, Sequence:, MAVAVRTLQEQLEKAKESLKNVDENIRKLTGRDPNDVRPIQARLLALSGPGGGRGRGSLLLRRGFSDSGGGPPAKQRDLEGAVSRLGGERRTRRESRQES
20 μL
Primary
Human, Rat
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
5411
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.