missing translation for 'onlineSavingsMsg'
Learn More

Pinacolborane, 97%, stabilized

Product Code. p-3737232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10738914 5 g Glass bottle 5g
10685483 25 g Glass bottle 25g
10571774 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10738914 Supplier Thermo Scientific Acros Supplier No. 371630050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25015-63-8 | C6H13BO2 | 127.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25015-63-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
MDL Number MFCD00674030
InChI Key UCFSYHMCKWNKAH-UHFFFAOYSA-N
Synonym pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n
PubChem CID 6364989
IUPAC Name 4,4,5,5-tetramethyl-1,3,2$l^{2}-dioxaborolane
SMILES CC1(C)OBOC1(C)C

Specifications

Color Colorless to Yellow
Density 0.8820g/mL
Boiling Point 42.0°C to 43.0°C (50.0 mmHg)
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3940 to 1.3980
Quantity 5 g
Specific Gravity 0.882
Stabilizer 0.6 to 1.4% triethylamine
Formula Weight 127.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pinacolborane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pinacolborane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.