missing translation for 'onlineSavingsMsg'
Learn More

Pinacol, 99%

Product Code. 11421607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421607 500 g 500g
11401607 25 g 25g
11411607 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421607 Supplier Thermo Scientific Alfa Aesar Supplier No. B25006.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-09-5 | C6H14O2 | 118.176 g/mol

Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.

Solubility
Soluble in hot water, alcohol, and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-09-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004462
InChI Key IVDFJHOHABJVEH-UHFFFAOYSA-N
Synonym pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
PubChem CID 6425
IUPAC Name 2,3-dimethylbutane-2,3-diol
SMILES CC(C)(C(C)(C)O)O

Specifications

Melting Point 42°C to 45°C
Boiling Point 171°C to 172°C
Flash Point 77°C (170°F)
Quantity 500 g
Beilstein 1340501
Sensitivity Hygroscopic
Merck Index 14,7439
Solubility Information Soluble in hot water,alcohol,and diethyl ether.
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material Pinacol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.