missing translation for 'onlineSavingsMsg'
Learn More

Pinacol, 99%

Product Code. 10137800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137800 250 g Glass bottle 250g
10772022 100 g Glass bottle 100g
11974631 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137800 Supplier Thermo Scientific Acros Supplier No. 418002500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pinacol, 99%, C6H14O2, CAS Number-76-09-5, pinacone, pinacol, unii-527qe7i5co, tetramethylethylene glycol, 2,3-dimethyl-2,3-butanediol, 2,3-dimethyl-2,3-dihydroxybutane, 1,1,2,2-tetramethylethylene glycol, 2,3-butanediol, 2,3-dimethyl, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-butane-2,3-diol | CAS: 76-09-5 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-09-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00004462
InChI Key IVDFJHOHABJVEH-UHFFFAOYSA-N
Synonym pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
PubChem CID 6425
IUPAC Name 2,3-dimethylbutane-2,3-diol
SMILES CC(C)(C(C)(C)O)O

Specifications

Melting Point 40.0°C to 43.0°C
Color White
Boiling Point 174.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2COHCOH(CH3)2
Quantity 250 g
Beilstein 01,487
Merck Index 15,755
Solubility Information Solubility in water: insoluble
Formula Weight 118.18
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Pinacol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Pinacol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.