Learn More
PIN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15789-1-AP-20UL
 View more versions of this product
Product Code. 16855124
  • £135.00 / 20µL
Description
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
Specifications
Specifications
PIN4
Polyclonal
Unconjugated
PIN4
EPVH, hEPVH, hPar14, hPar17, PAR14, PAR17, Parvulin 14, Parvulin 17, PIN4, PPIase Pin4, Rotamase Pin4
Rabbit
Antigen Affinity Chromatography
RUO
5303, 684441, 69713
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CWW6, Q9Y237
PIN4
PIN4 Fusion Protein Ag8489
20 μL
Primary
Human, Rat, Mouse