missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116804
This item is not returnable.
View return policy
Description
Positive Samples: 293T, HepG2, U-251MG, Mouse liver, Mouse brain, Rat testis, Rat brain Immunogen sequence: MPPKGKSGSG KAGKGGAASG SDSADKKAQG PKGGGNAVKV RHILCEKHGK IMEAMEKLKS GMRFNEVAAQ YSEDKARQGG DLGWMTRGSM VGPFQEAAFA LPVSGMDKPV FTDPPVKTKF GYHIIMVEGR K.
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
Specifications
| PIN4 | |
| Polyclonal | |
| Unconjugated | |
| Pin4 | |
| 2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5303, 684441, 69713 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.8 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9CWW6, Q9Y237 | |
| Pin4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-156 of human PIN4 (NP_0062142). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction