Learn More
Invitrogen™ PIN1 Monoclonal Antibody (5E5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549154
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Pin1 shares 95% amino acid (aa) sequence identity with mouse Pin1. Positive Control - WB: human Hela whole cell, human HEK293 whole cell, human SW620 whole cell, monkey COS-7 whole cell, rat brain tissue, rat C6 whole cell. ICC/IF: Hela cell. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.
Specifications
| PIN1 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q13526 | |
| Pin1 | |
| E.coli-derived human Pin1 recombinant protein (Position: M1-E163). | |
| 100 μg | |
| Primary | |
| Human, Monkey, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 5E5 | |
| Unconjugated | |
| Pin1 | |
| 0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 298696, 5300 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.