missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pin1 Antibody (3G8), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22956PECY55
This item is not returnable.
View return policy
Description
Pin1 Monoclonal specifically detects Pin1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Pin1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| PIN1 | |
| Recombinant human Pin1 (1-163 aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human Pin1 mAb, clone 3G8, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Pin1 protein. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 3G8 | |
| Flow Cytometry | |
| dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication, Mitotic Regulators, Phospho Specific | |
| 5300.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction