missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pin1 Antibody (3G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22956
This item is not returnable.
View return policy
Description
Pin1 Monoclonal specifically detects Pin1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, SDS-Page.
Specifications
| Pin1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Pin1 mAb, clone 3G8, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Pin1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, SDS-Page | |
| 3G8 | |
| Unconjugated | |
| NP_006212 | |
| PIN1 | |
| Recombinant human Pin1 (1-163 aa) purified from E. coli | |
| 0.1 ml | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 5300 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction