missing translation for 'onlineSavingsMsg'
Learn More

Pin1 Antibody (3G8), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18405133 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405133 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405133 Supplier Novus Biologicals Supplier No. NBP122956AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pin1 Monoclonal antibody specifically detects Pin1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pin1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 3G8
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Accession No. NP_006212
Gene Alias dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5
Host Species Mouse
Immunogen Recombinant human Pin1 (1-163 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5300
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.