missing translation for 'onlineSavingsMsg'
Learn More

PIN/DLC8 Rabbit anti-Human, Mouse, Rat, Clone: 3Q1J6, Novus Biologicals™

Product Code. 18339176 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339176 100 μg 100µL
18365135 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18339176 Supplier Novus Biologicals Supplier No. NBP316386100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PIN/DLC8 Monoclonal antibody specifically detects PIN/DLC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN/DLC8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3Q1J6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-89 of human PIN/DLC8 (P63167).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline p53 Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8655
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.