missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN/DLC8 Mouse anti-Human, Clone: 13C9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59394-100UL
This item is not returnable.
View return policy
Description
PIN/DLC8 Monoclonal specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PIN/DLC8 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1007, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 13C9 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| DYNLL1 | |
| Recombinant human DYNLL (1-89aa) purified from E. coli | |
| 100 μL | |
| p53 Pathway | |
| 8655 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction