missing translation for 'onlineSavingsMsg'
Learn More

PIN/DLC8 Antibody (13C9), DyLight 488, Novus Biologicals™

Product Code. 18431493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431493 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431493 Supplier Novus Biologicals Supplier No. NBP259394G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIN/DLC8 Monoclonal antibody specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN/DLC8
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 13C9
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase
Host Species Mouse
Immunogen Recombinant human PIN/DLC8 (1-89aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline p53 Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8655
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.