missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543252
This item is not returnable.
View return policy
Description
Peptide sequence: IDENPASDFD DSGSLLGFKY GSGQKYGGIP NFSHRQVRYL EKNVKLKSKY Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
Catalyzes the 2 serial methylation steps for the conversion of the 7-monomethylguanosine (m(7)G) caps of snRNAs and snoRNAs to a 2,2,7-trimethylguanosine (m(2,2,7)G) cap structure. The enzyme is specific for guanine, and N7 methylation must precede N2 methylation. Hypermethylation of the m7G cap of U snRNAs leads to their concentration in nuclear foci, their colocalization with coilin and the formation of canonical Cajal bodies (CBs). Plays a role in transcriptional regulation. [UniProt].
Specifications
| PIMT | |
| Polyclonal | |
| Unconjugated | |
| TGS1 | |
| Cap-specific guanine-N2 methyltransferase; CLL-associated antigen KW-2; D4Ertd800e; HCA137; Hepatocellular carcinoma-associated antigen 137; LOW QUALITY PROTEIN: trimethylguanosine synthase; NCOA6IP; nuclear receptor coactivator 6 interacting protein; nuclear receptor coactivator 6-interacting protein; Pimt; PIPMT; PRIP-interacting protein with methyltransferase motif; TGS1; tgs1 {ECO:0000250; Trimethylguanosine synthase; trimethylguanosine synthase 1; trimethylguanosine synthase homolog; trimethylguanosine synthase homolog (S. cerevisiae); trimethylguanosine synthase-like protein; UniProtKB:Q96RS0} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 96764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96RS0 | |
| TGS1 | |
| Synthetic peptide directed towards the middle region of human TGS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction