missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pimozide
GREENER_CHOICE

Product Code. 18732684
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18732684

Brand: Tocris Bioscience™ 0937/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D2-like antagonist

Dopamine D2-receptor antagonist and antipsychotic which binds with very high affinity to the cloned rat 5-HT7 receptor (Ki = 0.5 nM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 2062-78-4
Quantity 100 mg
Molecular Formula C28H29F2N3O
Formula Weight 461.55
Purity >99%
Molecular Weight (g/mol) 461.56
Chemical Name or Material Pimozide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.