missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pimozide

Product Code. 16514204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16514204 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16514204 Supplier Thermo Scientific Chemicals Supplier No. 242010010
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2062-78-4
Melting Point 214.0°C to 222.0°C
Molecular Formula C28H29F2N3O
MDL Number MFCD00055081
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in dimethylsulfoxide
InChI Key YVUQSNJEYSNKRX-UHFFFAOYSA-N
SMILES FC1=CC=C(C=C1)C(CCCN1CCC(CC1)N1C(=O)NC2=CC=CC=C12)C1=CC=C(F)C=C1
IUPAC Name 1-{1-[4,4-bis(4-fluorophenyl)butyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular Weight (g/mol) 461.56
Formula Weight 461.55
Chemical Name or Material Pimozide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.