missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pimonidazole
GREENER_CHOICE

Product Code. p-7116108
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18796406

Brand: Tocris Bioscience™ 6182/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hypoxia detection reagent

Pimonidazole is a hypoxia detection reagent. Labels hypoxic tumor cells in vivo. Also labels hypoxic mesenchymal stem cells in mouse bone marrow.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material α-[(2-Nitro-1H-imidazol-1-yl)methyl]-1-piperidineethanol
CAS 70132-50-2
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C11H18N4O3
Formula Weight Observed MW: 254.29
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.