missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pimonidazole

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Pimonidazole is a hypoxia detection reagent. Labels hypoxic tumor cells in vivo. Also labels hypoxic mesenchymal stem cells in mouse bone marrow.
Specifications
Specifications
| Chemical Name or Material | α-[(2-Nitro-1H-imidazol-1-yl)methyl]-1-piperidineethanol |
| CAS | 70132-50-2 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C11H18N4O3 |
| Formula Weight | Observed MW: 254.29 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction