missing translation for 'onlineSavingsMsg'
Learn More

Pimodivir, MedChemExpress

Product Code. 16443296
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16443296

Brand: Medchem Express HY12353A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit.

Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit.

Specifications

Chemical Name or Material Pimodivir
Physical Form Solid
CAS 1629869-44-8
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H19F2N5O2
For Use With (Application) COVID-19-anti-virus
Synonym VX-787
Solubility Information DMSO : 100 mg/mL (250.38 mM; Need ultrasonic)
SMILES O=C([C@H]1C(CC2)CCC2[C@@H]1NC3=NC(C4=CNC5=NC=C(F)C=C54)=NC=C3F)O
Molecular Weight (g/mol) 399.39
Formula Weight 399.39
Percent Purity 98.17%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.