missing translation for 'onlineSavingsMsg'
Learn More

Pimethixene maleate, MedChemExpress

Product Code. 16438668
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438668 Supplier Medchem Express Supplier No. HYB1101A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pimethixene maleate is antihistamine and antiserotonergic compound, acts as an antimigraine agent. Pimethixene maleate is a highly potent antagonist of 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4 as well as muscarinic M1 and M2 receptors, with pKis of 7.63, 10.22, 10.44, 8.42, 10.14, 8.19, 7.54, 8.61 and 9.38, respectively.

Pimethixene maleate is antihistamine and antiserotonergic compound, acts as an antimigraine agent.

Specifications

Chemical Name or Material Pimethixene maleate
CAS 13187-06-9
Color White
Purity Grade Notes Research
Molecular Formula C23H23NO4S
Quantity 10 mg
Synonym Pimetixene maleate
Solubility Information DMSO : 100 mg/mL (244.20 mM; Need ultrasonic)
SMILES CN1CC/C(CC1)=C2C3=C(SC4=C\2C=CC=C4)C=CC=C3.OC(/C=C\C(O)=O)=O
Molecular Weight (g/mol) 409.5
Formula Weight 409.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.