missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-20 R beta Protein, Fc Tag

Product Code. 16417749
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg

Product Code. 16417749

Brand: ACROBiosystems ILBH52581mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Interleukin-20 receptor subunit beta (IL20RB) is also known as IL-20R2, DIRS1, which belongs to the type II cytokine receptor family. IL20RB / IL-20R2 contains two fibronectin type-III domains. IL20RB / IL-20R2 is widely expressed with highest levels in skin and testis. IL20RB can be fused with IL20RA or IL22RA1 forming heterodimer. The IL20RA/IL20RB dimer is a receptor for IL19, IL20 and IL24. The IL22RA1/IL20RB dimer is a receptor for IL20 and IL24.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.1 kDa. As a result of glycosylation, the protein migrates as 50 kDa under reducing (R) condition, and 100 kDa under non-reducing (NR) condition (SDS-PAGE).

Specifications

Accession Number AAH63696
Molecular Weight (g/mol) 49.1 kDa
Quantity 1 mg
Source Human IL-20 R beta, Fc Tag (ILB-H5258) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Ala 230 (Accession # AAH63696).
Immunogen Asp 30 - Ala 230
Storage Requirements -20°C
Gene Alias IL20RB,IL-20R2,IL-20 R-beta,DIRS1,FNDC6
Common Name IL-20 R beta
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer 50 mM Tris, 100 mM Glycine, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Other Recombinant Proteins
Purity or Quality Grade >90% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.