missing translation for 'onlineSavingsMsg'
Learn More

Pimelic Acid, TRC

Product Code. 30138261
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138261 50 g 50g
30132529 100 g 100g
30011573 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138261 Supplier Toronto Research Chemicals Supplier No. TRCP44504050G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-16-0
Molecular Formula C7 H12 O4
Molecular Weight (g/mol) 160.17
Synonym Heptanedioic acid, Pimelic acid (8CI), 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid, NSC 30112, Pileric acid, Pimeric acid
IUPAC Name heptanedioic acid
SMILES OC(=O)CCCCCC(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H12O4/c8-6(9)4-2-1-3-5-7(10)11/h1-5H2,(H,8,9)(H,10,11)
Formula Weight 160.0736
Chemical Name or Material 1,5-Pentanedicarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.