missing translation for 'onlineSavingsMsg'
Learn More

Pimelic acid, 98%

Product Code. 11488407
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488407 100 g 100g
11478407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488407 Supplier Thermo Scientific Alfa Aesar Supplier No. A18495.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-16-0 | C7H12O4 | 160.17 g/mol

Pimelic acid derivatives are involved in the biosynthesis of the amino acid called lysine. It is applied as a precursor to many polyesters and polyamides. It is the final member of the mnemonic used to aid recollection of the order of the first six dicarboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pimelic acid derivatives are involved in the biosynthesis of the amino acid called lysine. It is applied as a precursor to many polyesters and polyamides. It is the final member of the mnemonic used to aid recollection of the order of the first six dicarboxylic acids.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-16-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004425
InChI Key WLJVNTCWHIRURA-UHFFFAOYSA-N
Synonym pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f
PubChem CID 385
ChEBI CHEBI:30531
IUPAC Name heptanedioic acid
SMILES OC(=O)CCCCCC(O)=O

Specifications

Melting Point 102°C to 106°C
Density 1.329
Boiling Point 212°C (10 mmHg)
Linear Formula HO2C(CH2)5CO2H
Quantity 100 g
Beilstein 1210024
Merck Index 14,7431
Solubility Information Soluble in water. Solubility in methanol is almost transparent.
Formula Weight 160.17
Percent Purity ≥98%
Chemical Name or Material Pimelic acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.