missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pimavanserin

Click to view available options
:
50 mg
Description
Pimavanserin (also known as ACP-103) is a selective, potent 5-HT2A receptor inverse agonist (IC50 = 2 nM). Pimavanserin displays ∼ 40-fold higher affinity for 5-HT2A over 5-HT2C receptor. Chronic administration of Pimavanserin, suppresses amyloid-β production and improves anxiety-related behavior and memory in Alzheimer's disease mouse models. Pimavanserin shows protective effects on midbrain dopaminergic neurons in a rat model of Parkinson's Disease.
Specifications
Specifications
| Chemical Name or Material | N-[(4-Fluorophenyl)methyl]-N-(1-methyl-4-piperidinyl)-N'-[[4-(2-methylpropoxy)phenyl]methyl]urea |
| CAS | 706779-91-1 |
| Quantity | 50 mg |
| Target | 5-HT2A Receptor Antagonists |
| Molecular Formula | C25H34FN3O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction