missing translation for 'onlineSavingsMsg'
Learn More

Pimavanserin, MedChemExpress

Product Code. 16426496
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16426496 100 mg 100mg
16412237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16426496 Supplier Medchem Express Supplier No. HY14557
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pimavanserin is a selective inverse agonist of the 5-HT2A receptor with pIC50 and pKd of 8.73 and 9.3, respectively.

Pimavanserin is a selective inverse agonist of the 5-HT2A receptor with pIC50 and pKd of 8.73 and 9.3, respectively.

Specifications

Chemical Name or Material Pimavanserin
CAS 706779-91-1
Color White
Purity Grade Notes Research
Molecular Formula C25H34FN3O2
Quantity 100 mg
Synonym ACP-103
Solubility Information DMSO : 50 mg/mL (116.95 mM; Need ultrasonic)
SMILES CC(C)COC1=CC=C(CNC(N(CC2=CC=C(F)C=C2)C3CCN(C)CC3)=O)C=C1
Molecular Weight (g/mol) 427.55
Formula Weight 427.55
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.