missing translation for 'onlineSavingsMsg'
Learn More

Pimasertib, MedChemExpress

Product Code. 16447687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447687 5 mg 5mg
16444948 10 mg 10mg
16425726 100 mg 100mg
16411197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447687 Supplier Medchem Express Supplier No. HY120425mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pimasertib (AS703026) is a highly selective, ATP non-competitive allosteric orally available MEK1/2 inhibitor.

Pimasertib (AS703026) is a highly selective, ATP non-competitive allosteric orally available MEK1/2 inhibitor.

Specifications

Chemical Name or Material Pimasertib
CAS 1236699-92-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C15H15FIN3O3
Quantity 5 mg
Synonym AS703026 MSC1936369B
Solubility Information DMSO : ≥ 100 mg/mL (231.91 mM)
SMILES FC1=C(C=CC(I)=C1)NC2=CN=CC=C2C(NC[C@@H](CO)O)=O
Molecular Weight (g/mol) 431.2
Formula Weight 431.2
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.