missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIM2 Recombinant Human Protein
PIM2 is a proto-oncogene.
Brand: Invitrogen™ PV3649
Code : 50
Additional Details : Weight : 0.00945kg
Description
PIM2 phosphorylates BAD, which partially inhibits its apopotopic function.
Specifications
NP_006866 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
63.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length, Recombinant | |
pim-2 oncogene | |
Human | |
GST Tag | |
Kinases & Inhibitors |
Apoptosis, Apoptosis, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Industrial and Applied Science, Kinase Assay, Cell Viability, Kinase Proteins and Expression Reagents, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma | |
11040 | |
10μg | |
RUO - research use only | |
Pim2 | |
PIM Kinase Family | |
Recombinant | |
Baculovirus | |
Serine/Threonine Kinases |