missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25865-1-AP-20UL
This item is not returnable.
View return policy
Description
PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.Specifications
| PIM2 | |
| Polyclonal | |
| Unconjugated | |
| PIM2 | |
| pim 2 oncogene, Pim 2h, PIM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11040 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P1W9 | |
| PIM2 | |
| PIM2 Fusion Protein Ag23130 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction