missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Rabbit anti-Human, Clone: 3V1S5, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | PIM2 |
|---|---|
| Clone | 3V1S5 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18355064
|
Novus Biologicals
NBP3-16789-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365534
|
Novus Biologicals
NBP3-16789-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIM2 Monoclonal antibody specifically detects PIM2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| PIM2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 11040 | |
| IgG | |
| Affinity purified |
| 3V1S5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11.1, pim-2 oncogene, pim-2h, proto-oncogene Pim-2 (serine threonine kinase), serine/threonine protein kinase pim-2, serine/threonine-protein kinase pim-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-120 of human PIM2 (Q9P1W9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title