missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM2 Polyclonal Antibody

PIM2 Polyclonal Antibody
Brand: Invitrogen PA5142734
This item is not returnable.
View return policy
Description
PIM2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| PIM2 | |
| Polyclonal | |
| Unconjugated | |
| PIM2 | |
| DXCch3; PIM2; Pim-2; pim-2 oncogene; Pim-2 proto-oncogene, serine/threonine kinase; pim-2h; proto-oncogene Pim-2 (serine threonine kinase); proviral integration site 2; serine/threonine protein kinase pim-2; serine/threonine-protein kinase pim-2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 11040 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9P1W9 | |
| PIM2 | |
| Peptide with sequence C-KDREAFEAEYRLGP (aa 23-36). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction